Learn More
Boric Acid, Powder, NF, 50 kg, Spectrum Chemical
SDP
Catalog No. 18615444
Click to view available options
:
Quantity: 50 kg
Boric Acid, Powder, NF is used as an antimicrobial preservative, a buffering agent and as a tonicity adjusting excipient. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
Quantity 50 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.