Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Acid, Purified, Reagents

Catalog No. 01337608
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-608 500 g
01-337-609 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-608 Supplier Reagents Holdings Llc Supplier No. C2104300500A5
Add to Cart
Add to Cart

Boric Acid, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym Orthoboric Acid, Boracic Acid, Borofax, Trihydroxyborane
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Colorless/White
Melting Point 168°C to 170°C
Boiling Point 300°C
Packaging Poly Bottle
Specific Gravity 1.435
Quantity 500 g
Physical Form Solid
Grade Pure
Chemical Name or Material Boric Acid, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.