Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boric Anhydride, Purified, J.T.Baker™

Click to view available options
Quantity:
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

1303-86-2
B2O3
69.617
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
518682
CHEBI:30163
CAS 1303-86-2
Packaging Poly Bottle
Specific Gravity 1L = 1.85kg
Molecular Formula B2O3
Quantity 2.5kg
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
SMILES B(=O)OB=O
IUPAC Name oxo(oxoboranyloxy)borane
Molecular Weight (g/mol) 69.617
PubChem CID 518682
ChEBI CHEBI:30163
Formula Weight 69.62
Chemical Name or Material Boric Anhydride
Grade Purified
Show More Show Less