Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boris / CTCFL Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992903
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2903 100 μL
50-199-2904 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2903 Supplier Active Motif Supplier No. 39851
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.

Specifications

Antigen Boris / CTCFL
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene Ctcfl
Gene Accession No. Q8NI51
Gene Alias BORIS; BORIS like protein; BORIS-like protein; Brother of the regulator of imprinted sites; Cancer/testis antigen 27; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein)-like; CCCTC-binding factor like; CT27; CTCF paralog; CTCF T; Ctcfl; CTCF-like protein; CTCF-T; dJ579F20.2; HMG 1L1; HMG-1L1; HMGB1L1; likely orthologue of H. sapiens CCCTC-binding factor-like protein (CTCFL); MGC163358; MGC169105; MGC169106; OTTMUSG00000016680; putative high mobility group protein 1-like 1; putative high mobility group protein B1-like 1; Transcriptional repressor CTCFL; Zinc finger protein CTCF-T
Gene Symbols Ctcfl
Host Species Rabbit
Immunogen This antibody was raised against a peptide within the N-terminal region of human BORIS.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.