Learn More
BORISRabbit anti-Chimpanzee, Human, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401907

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against the human BORIS protein. A BLAST analysis was used to suggest cross reactivity with BORIS proteins from human and chimpanzee. Partial reactivity may occur against BORIS from mouse, dog and rat sources based on varying degrees of homology to the immunizing sequence. Reactivity against homologues from other sources is not known.
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.Specifications
BORIS | |
Polyclonal | |
Unconjugated | |
Ctcfl | |
BORIS; BORIS like protein; BORIS-like protein; Brother of the regulator of imprinted sites; Cancer/testis antigen 27; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein)-like; CCCTC-binding factor like; CT27; CTCF paralog; CTCF T; Ctcfl; CTCF-like protein; CTCF-T; dJ579F20.2; HMG 1L1; HMG-1L1; HMGB1L1; likely orthologue of H. sapiens CCCTC-binding factor-like protein (CTCFL); MGC163358; MGC169105; MGC169106; OTTMUSG00000016680; putative high mobility group protein 1-like 1; putative high mobility group protein B1-like 1; Transcriptional repressor CTCFL; Zinc finger protein CTCF-T | |
Rabbit | |
RUO | |
140690, 458360 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.28 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Q8NI51 | |
Ctcfl | |
100 μg | |
Primary | |
Chimpanzee, Human | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.