Learn More
Boron ICP Standard, 1000 ppm B in H2O/tr NH4OH, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 97.0, 0.56, 0.1 |
| CAS Max % | 100.0, 0.58, 0.1 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Type | Standard |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | 1mL = 1mg B (1, 000ppm B)H3BO3 in H2O / trace NH4OH |
| Chemical Name or Material | Boron ICP Standard |
Safety and Handling
| ShelfLife | 18 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.