Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron, plasma standard solution, Specpure™ B 1000μg/mL

Catalog No. AA13859AE
Click to view available options
Quantity:
50 mL
100 mL
500 mL

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00134034

Specifications

Physical Form Liquid
Concentration H3BO3 in 1% NH4OH
Quantity 100 mL
Grade Specpure™
Chemical Name or Material Boron plasma standard solution, B 1000μg/mL
Hazard Category H315-H319-H360FD
Hazard Statement GHS H Statement
H360-H315-H319
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.