Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. SB155100 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle

For atomic absorption. (1mL = 1mg B) Solute: Boric Acid Solvent: Distilled Water

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3, 7732-18-5
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Packaging Poly Bottle
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Physical Form Liquid
Concentration 990 to 1010 ppm
Color Colorless
Melting Point 0°C
Quantity 100 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Boron, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.