Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron Trifluoride, 12% (1.5 M) in Methanol, Thermo Scientific Chemicals™ PROMO

Catalog No. AC402765S24
Click to view available options
:
Boron Trifluoride, 12% (1.5 M) in Methanol; Packaging: Glass Bottle; Quantity: 500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Synonym: Boron Trifluoride Methanol, BF3 Methanol, Borontrifluoride Methanol

  • Originally part of the Acros Organics™ product portfolio
  • Some documentation and label information may refer to the legacy brand
  • Original Acros Organics™ product and item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals™
  • Acylation reagent and Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373-57-9, 67-56-1
Molecular Formula BF3
Molecular Weight (g/mol) 67.81
MDL Number MFCD00011316
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
Synonym boron trifluoride-methanol solution
PubChem CID 11062313
IUPAC Name trifluoroborane
SMILES FB(F)F

Specifications

Boiling Point 65°C
Melting Point -98°C
Color Amber to Colorless
Physical Form Liquid
Quantity 500 g
Linear Formula BF3
Flash Point 4°C
Merck Index 14, 1349
Solubility Information Soluble in Water (May Decompose)
Formula Weight 67.81
Percent Purity 10 to 15%
Packaging Glass Bottle
Specific Gravity 0.87
Density 0.87 g/mL
Chemical Name or Material Boron Trifluoride
Show More Show Less
Hazard Statement Reacts violently with water.
Precautionary Statement Combustible liquid. Causes severe skin burns and eye damage. Fatal if inhaled. Toxic if swallowed.
EINECSNumber 206-766-4
RTECSNumber ED2275000
TSCA TSCA

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.