Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boron Trifluoride, 14% in Methanol, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Boron Trifluoride, 14 Percent in Methanol is used as a reagent in organic chemistry most often as a Lewis acid. It is known to be a versatile building block in several other boron compounds. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
CAS Max % | 100% |
Quantity | 500 mL |
UN Number | UN3286 |
Grade | Reagent |
Packaging | Amber Glass Bottle |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction