Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Boron trifluoride tetrahydrofuran complex 50%, Thermo Scientific™
Click to view available options
500mL; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acid
Specifications
CAS | 462-34-0 |
Quantity | 500mL |
Assay Percent Range | 50% |
Molecular Formula | C4H8BF3O |
MDL Number | MFCD00040372 |
Synonym | boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor |
InChI Key | CHNLPLHJUPMEOI-UHFFFAOYSA-N |
SMILES | FB(F)F.C1CCOC1 |
IUPAC Name | oxolane; trifluoroborane |
Molecular Weight (g/mol) | 139.91 |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS P Statement Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Harmful if inhaled. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction