Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Borosilicate Glass Test Tubes with Rim
SDPSCI_ED

Catalog No. S15930 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15930 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15930 Supplier United Scientific™ Supplier No. TT9800K
Add to Cart
Add to Cart

Safely packaged test tubes with good clarity

  • Pre-cleaned borosilicate glass test tubes with rims.
  • Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.

Specifications

Capacity (Metric) 175 mm
Length (Metric) 200 mm
Material Borosilicate Glass
Shape Cylindrical
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No
Product Type Reusable Test Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.