Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borrelia Burgdorferi Antibody (BGN/F44/42), HRP

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066394H
Description
Borrelia Burgdorferi Monoclonal specifically detects Borrelia Burgdorferi in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Borrelia Burgdorferi | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein A purified | |
RUO | |
NB100-66394 recognizes Borrelia burgdorferi, a gram-negative spirochete that is a causative agent of lyme disease in humans. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA | |
BGN/F44/42 | |
Western Blot, ELISA | |
B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease | |
Native | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction