Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borrelia Burgdorferi Flagellar Antigen Antibody (7G11/6802), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11059982AF594
Description
Borrelia Burgdorferi Flagellar Antigen Monoclonal specifically detects Borrelia Burgdorferi Flagellar Antigen in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Borrelia Burgdorferi Flagellar Antigen | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
Reacts with a flagellar antigen. | |
Store at 4°C in the dark. |
ELISA, Immunocytochemistry, Immunofluorescence | |
7G11/6802 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease | |
Native | |
Primary | |
Bacteria | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction