Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Borrelia burgdorferi Monoclonal Antibody (D36E)
GREENER_CHOICE

Catalog No. PIMA17284
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17284 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17284 Supplier Invitrogen™ Supplier No. MA17284
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7284 antibody reacts with Borrelia burgdorferi Osp B in bacterial and infected tissue samples. MA1-7284 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7284 antibody was raised against RV Osp B antigen.

Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Borrelia burgdorferi
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone D36E
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias B. burgdorferi; Borrelia; Lyme disease
Host Species Mouse
Immunogen RV Osp B antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.