Learn More
Invitrogen™ Borrelia burgdorferi Polyclonal Antibody, Biotin

Description
The PA1-73007 antibody reacts with Borrelia burgdorferi. PA1-73007 has been successfully used in Immunofluorescence applications. The PA1-73007 antibody was raised against B. burgdorferi whole cell preparation.
Specifications
Specifications
| Antigen | Borrelia burgdorferi |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | B. burgdorferi; Borrelia; Lyme disease |
| Host Species | Rabbit |
| Immunogen | B. burgdorferi whole cell preparation. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.