Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Borrelia burgdorferi Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PA173006
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173006 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173006 Supplier Invitrogen™ Supplier No. PA173006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73006 antibody reacts with Borrelia burgdorferi. Some cross-reactivity with Treponema pallidum, B. hermsii, and B. parkerii. PA1-73006 has been successfully used in ELISA applications. The PA1-73006 antibody was raised against B. burgdorferi whole cell preparation.

Borrelia burgdorferi is a spirochete and the cause of Lyme disease, a tick transmitted illness of humans and animals. B. burgdorferi may persist in humans and animals for months or years following initial infection, despite a robust humoral immune response. B. burgdorferi resembles other spirochetes in that it is a highly specialized, motile, two-membrane, spiral shaped bacteria which lives primarily as an extracellular pathogen. B. burgdorferi has an unusual genome compared with other eubacteria which includes a linear chromosome approximately one megabase in size and numerous linear and circular plasmids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Borrelia burgdorferi
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal
Gene Alias B. burgdorferi; Borrelia; Lyme disease
Host Species Rabbit
Immunogen B. burgdorferi whole cell preparation.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.