Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bosentan
GREENER_CHOICESDP

Catalog No. 623250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-325-0 50 mg
62-321-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-325-0 Supplier Tocris Bioscience™ Supplier No. 6232/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High affinity dual ETA and ETB receptor antagonist;orally bioavailable

Bosentan is a high affinity dual ETA and ETB receptor antagonist (Ki values are 4.7 and 95 nM for ETA and ETB, respectively). Inhibits ET-1 induced contraction of isolated rat aorta and sarafotoxin S6C -induced contraction of rat trachea in vitro. Orally bioavailable.

Specifications

CAS 147536-97-8
Quantity 50 mg
Target Non-selective Endothelin Receptor
Molecular Formula C27H29N5O6S
Formula Weight 551.61g/mol
Product Type Antagonist

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.