Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, 500mL
SDP

Catalog No. p-7111692 Shop All DWK Life Sciences Products
Click to view available options
Closure Liner:
Polyethylene
Polyethylene Cone
Closure Material:
Phenolic
Polypropylene

Wheaton clear safety coated Boston Round bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.

  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Coated glass bottles provide an added measure of safety
  • Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Coating Safety Coated
Color Clear
Material Borosilicate Glass (Type 3)
Quantity 12/Case
Closure Color White
Closure Included Yes
Mouth Wide
Product Type Boston Round Bottle
Closure Liner Polyethylene
Closure Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.