Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bosutinib

Catalog No. AC465650010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465650010 1 g
AC465652500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465650010 Supplier Thermo Scientific Chemicals Supplier No. 465650010
Add to Cart

CAS: 380843-75-4 | C26H29Cl2N5O3 | 530.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380843-75-4
Molecular Formula C26H29Cl2N5O3
Molecular Weight (g/mol) 530.45
MDL Number MFCD07367846
InChI Key UBPYILGKFZZVDX-UHFFFAOYSA-N
IUPAC Name 4-[(2,4-dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methylpiperazin-1-yl)propoxy]quinoline-3-carbonitrile
SMILES COC1=CC(NC2=C3C=C(OC)C(OCCCN4CCN(C)CC4)=CC3=NC=C2C#N)=C(Cl)C=C1Cl

Specifications

Melting Point 131.0°C to 134.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol and dmso
Formula Weight 530.45
Chemical Name or Material Bosutinib
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.