Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bosutinib
GREENER_CHOICE

Catalog No. 436110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-611-0 10 mg
43-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-611-0 Supplier Tocris Bioscience™ Supplier No. 4361/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dual Src-Abl inhibitor; antiproliferative

Dual inhibitor of Abl and Src kinases (IC50 = 1.2 nM for Src in an enzymatic assay). Displays antiproliferative activity against chronic myelogenous leukemia (CML) cells and decreases the motility and invasion of breast cancer cell lines. Also exhibits potent antiproliferative activity in anchorage-independent, Src-transformed rat fibroblasts (IC50 = 100 nM). Displays selectivity for Src over non-Src family kinases such as growth factor receptor tyrosine kinases.

Chemical Identifiers

CAS 380843-75-4
Molecular Formula C26H29Cl2N5O3

Specifications

Quantity 10 mg
Formula Weight 530.45
Purity >99%
Molecular Weight (g/mol) 530.45
Chemical Name or Material Bosutinib
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.