Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Bottle Kit
Shop All Siemens Healthineers Products
Click to view available options
Capacity (Metric):
5 mL
15 mL
Quantity:
10 x 5 mL
10 x 15 mL
Specifications
Specifications
| For Use With (Equipment) | Coagulation Analyzers |
| Capacity (Metric) | 5 mL |
| Quantity | 10 x 5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction