Learn More
Eppendorf™ Bottle Thread Adapter
Catalog No. 05414299 Shop All Eppendorf North America Products
Change view
Click to view available options
Thread Size:
GL 32 to GL/S 28
Catalog No. Thread Size
05-414-299 GL 32 to GL/S 28
1 options
This item is not returnable. View return policy
Catalog No. 05-414-299 Supplier Eppendorf™ Supplier No. 4960835005
Add to Cart
Add to Cart
This item is not returnable. View return policy
Bottle Thread Adapter from GL 32 to GL/S 28
  • ETFE
  • From GL 32 to GL/S 28
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bottle Thread Adapter
Material ETFE
Thread Size GL 32 to GL/S 28