Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Bottle Top Dispenser with Bottle
SDPSCI_ED

Catalog No. p-7229329 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
0.25 to 2.5 mL
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 30 mL
5 to 60 mL
Increments:
0.05 mL
0.1 mL
0.2 mL
0.5 mL
1 mL
2 mL
Weight (English):
1.5 lb.
1.75 lb.
2 lb.
2.25 lb.
4 lb.
Volume (Metric) Reservoir:
100 mL
250 mL
500 mL
1000 mL
2000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Increments Weight (English) Volume (Metric) Reservoir
S139567 0.25 to 2.5 mL 0.05 mL 1.5 lb. 100 mL
S139568 0.5 to 5 mL 0.1 mL 1.5 lb. 100 mL
S139569 1 to 10 mL 0.2 mL 1.75 lb. 250 mL
S139570 2.5 to 30 mL 0.5 mL 2 lb. 500 mL
S139571 5 to 60 mL 1 mL 2.25 lb. 1000 mL
S139572 10 to 100 mL 2 mL 4 lb. 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. S139567 Supplier United Scientific™ Supplier No. BTDR1SET
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Convenient kit which includes a bottle top dispenser and GL45 Media Bottle.

United brand bottle top dispensers offer high accuracy at economical prices.

  • Each dispenser is individually calibrated and certified in an ISO 17025 accredited laboratory in accordance with ISO 8655 standards
  • A set of adapters to fit most laboratory reagent bottles are provided
  • All wetted parts are made from PTFE and borosilicate glass for maximum chemical resistance
  • These easy-to-use instruments are suitable for all laboratory applications, except HF
  • Special precautions must be taken for use, cleaning, and storage when working with concentrated fuming acids
  • A factory calibration certificate is provided with each bottle top dispenser
  • Dispensers are easy to disassemble and can be recalibrated in the lab
  • Fully autoclavable at 121°C, 15 psi for 10 to 15 minutes

Specifications

Accuracy 0.006
Product Type Analog Bottletop Dispenser with Adapters
Autoclavable Autoclavable
Certifications/Compliance ISO 8655
Color White
Channels 1
Material PTFE, Borosilicate Glass
Volume (Metric) 0.25 to 2.5 mL
Display Analog
For Use With (Application) Dispensing Liquids
For Use With (Equipment) Chemicals
Includes Dispenser and Glass Bottle
Increments 0.05 mL
Sterility Non-sterile
Volume (Metric) Reservoir 100 mL
Warranty 1 Year
Weight (English) 1.5 lb.
No. per Pack 1/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.