Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Bottle Top Dispenser with Dual Inlet Technology

Catalog No. 13681533 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
0.5 to 5 mL
1 to 10 mL
10 to 100 mL
2.5 to 30 mL
5 to 60 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-681-533 2.5 to 30 mL
13-681-531 0.5 to 5 mL
13-681-532 1 to 10 mL
13-681-534 5 to 60 mL
13-681-535 10 to 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 13-681-533
Add to Cart
Add to Cart
This item is not returnable. View return policy

Dual Inlet Technology facilitates to re-fill the bottle and rinse the dispenser. Bottle Top Dispenser with Dual Inlet Technology provides greater chemical compatibility.

  • Dual Inlet Technology facilitates to re-fill the bottle, rinse the dispenser, perform dilution and dual reagent dispensing without dis-mounting the dispenser from its bottle
  • Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and no maintenence required
  • Presence of a unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, also ensures wiper like smooth and effortless piston movement
  • Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
  • Adjustable delivery nozzle provides flexibility to the users in demanding laboratory conditions
  • Bottle neck adapters of different sizes (28, 30, 32, 36, 40 and 45mm) ensures compatibility of dispenser with majority of reagent bottles
  • Each dispenser is accompanied with tool, which helps in easy and quick re-calibration
  • Contemporary and well designed contours, silky surface finish
  • Individually calibrated as per ISO 8655-2

Specifications

Product Type Bottletop Dispenser
Autoclavable Only the piston needs to be removed for autoclaving the instrument. Piston is also autoclavable
Certifications/Compliance CE Certificate, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655, Calibration lab certification by NABL as per ISO 17025
Color White and Fisher Blue
Density 2.2 g/cm3
Description Shape: Cylindrical
Material Polypropylene - Outer Body (10% Glass Filled)
Volume (Metric) 2.5 to 30 mL
For Use With (Equipment) Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel.
Includes Calibration tool, Adapters set (4 nos.), Instruction Manual 4, Inlet tube, Calibration report A, Calibration report B, Inlet tube for the second inlet, Caps for the second bottle (4 nos.)
Vapor Pressure 300mbar to 600mbar
Viscosity Rating 500mm2
Warranty 1 year free replacement warranty against any manufacturing defects.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.