Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Bottle Top Dispensers (BTD) Cylinders

Catalog No. 10320156 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
10-320-156 10 mL
10-320-154 2.5 mL
10-320-155 5 mL
10-320-157 25 mL
10-320-158 50 mL
10-320-159 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 10-320-156 Supplier Corning™ Supplier No. 6855
Add to Cart
Add to Cart

Dispenses reagent to avoid losing solution during normal transfer


  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 10 mL
    For Use With (Application) Dispensing Organic and Inorganic Liquids
    Includes Four adapters
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.