Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bottle Top Valves and Filters
SDP

Catalog No. 06611821 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Inlet Valve Without Filter
Inlet Valve/Filter (15 mm)
Inlet Valve/Filter (4 mm)
Outlet Valve/Filter (4 mm)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
06-611-821 Inlet Valve/Filter (15 mm)
06-611-820 Inlet Valve/Filter (4 mm)
06-611-819 Inlet Valve Without Filter
06-611-822 Outlet Valve/Filter (4 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-611-821 Supplier Restek™ Supplier No. 27840
Add to Cart
Add to Cart

Protect labs from volatile compounds; use with a filter to also protect mobile phases from bacterial contamination

  • Outlet valve regulates pressure during helium sparging
  • 1/4-28 threads for use on Mobi-Cap or Hub-Cap Bottle Tops
  • Replace valves every 6 months and filters every 3 months; more frequently with rigorous use or use in harsh environments

Specifications

For Use With (Application) Bottle Cap
Quantity 1
Sterility Non-sterile
Thread Size (Metric) 1/4-28
Neck Finish 78 mm
Product Type Inlet Valve/Filter (15 mm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.