Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Bottled Media for Sartorius Membrane Filtration Units

Catalog No. 5090117905 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Application):
Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms
Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms
Membrane filtration method or for direct incubation, for determination of Yeasts and molds
Membrane filtration method or for direct incubation, for determination of total count
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
5090117905 Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms
5090117907 Membrane filtration method or for direct incubation, for determination of Yeasts and molds
5090117904 Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms
5090117903 Membrane filtration method or for direct incubation, for determination of total count
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 5090117905 Supplier Sartorius Supplier No. 14155A

Lysine agar media for wild yeasts determination

The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.

  • 250mL bottles, 4 bottles per box
  • Economical, provides time and cost savings
  • According to international formulas
  • Consistent quality
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Format Bottle
Manufacturing Origin Germany
Media Type Lactose
Product Type Broth
Sterility Sterile
Target Organism Class Coliforms
Test Type Microbial Detection
Fill Volume 250 mL
For Use With (Application) Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms
For Use With (Equipment) Sartorius Membrane Filtration Units
Incubation Conditions 35°C to °C;
Incubation Time 18 to 48 hr.
Packaging Type Bottle
Quantity 250 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.