Learn More
Sigma Aldrich Bottom Perforated Petri Dish For Live Cell Fluorescence Imaging
Catalog No. 503125538 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
50-312-5538 1 Pk.
1 options
Catalog No. 50-312-5538 Supplier Sigma Aldrich Supplier No. TEM3755
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Bottom perforated petri dish for loading Cry0Capsule for direct fliorescence imaging of living cells
Specifications
Quantity 1 Pk.