Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Bottom Sampling Dredge
SDPSCI_ED

Catalog No. s67778 Shop All LaMotte Company Products
Click to view available options
Material:
Stainless Steel

Collect river bottom samples easily.

  • Stainless steel
  • Trigger holds the sampler open while lowering
  • Scissor design closes sampler securely
  • Volume: 67 cu. in.
  • For soft bottoms (sand or silt)

Specifications

Grade 3+
Product Type Bottom Sampling Net
For Use With (Application) Environmental education
Material Stainless Steel
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.