Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGuard™ Bouffant Caps
Click to view available options
Style:
Pancake
Pleated
Color:
Green
Red
White
Description
- Strong, elastic headband
- Superior barrier properties
- Available in pancake or pleated styles
Specifications
Specifications
| Material | Spunbond Polypropylene |
| Disposable | Single-use |
| Model | B11A22 |
| For Use With (Application) | Food Service, Food Processing, Industrial, Janitorial, Manufacturing |
| Style | Pancake |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction