Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
StatLab™ Bouin's 2000 Fixative

Click to view available options
Quantity:
1 L
1 pt.
100 mL
4 gal./Cs.
Description
- Low-hazardous replacement for Bouin's Fluid
- Soft and delicate structures are preserved and trichrome stains will be brilliant with color while demonstrating crisp nuclear detail
- An ideal fixative to use on biopsy specimens of the gastrointestinal tract because the nuclear staining performs better than 10% formalin
Specifications
Specifications
Color | Yellow |
For Use With (Equipment) | All Tissue Processors |
Grade | Chemical Solution |
Product Type | Fixative |
Quantity | 4 gal./Cs. |
For Use With (Application) | Fixation |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction