Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOULE Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982443
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2443 Supplier Bioss Supplier No. BS12880R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen BOULE
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BOLL
Gene Accession No. Q8N9W6
Gene Alias 4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); BOLL; BOS_1970; Boule; boule homolog, RNA binding protein; boule-like; boule-like RNA binding protein; boule-like RNA-binding protein; LOC100341687; Protein boule-like; RGD1559527
Gene Symbols BOLL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BOULE.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66037
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.