Description
- Lightweight, rimless design offers optimal protection
- Flexible bayonet style temples provide a secure fit
- Molded nose bridge design to fit the vast majority of users
- Light Blue Lens - visible light transmission of 58% which relieves eye strain from bright, overhead fluorescent lighting
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
- Anti-Fog Coating eliminates the condensation of moisture on lenses that causes fogging so your lenses and vision stay clear
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Light Blue |
| Product Type | Safety glasses |
| Frame Color | Light Blue |
| Lens Material | Polycarbonate |