Description
- Lightweight, rimless design offers optimal protection
- Flexible bayonet style temples provide a secure fit
- Molded nose bridge design to fit the vast majority of users
- Clear Lens - clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
- Anti-Reflective (AR) lens coating will reduce reflection and glare, which can be particularly noticeable when working indoors, or in conditions with multiple light sources such as night driving, quality control and inspection, and repair/maintenance
- Treated front and back with a clear, scratch-resistant coating for a much harder surface that is more resistant to scratching
Specifications
Specifications
| Lens Coating | Anti-reflective, Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Safety glasses |
| Frame Color | Clear |
| Lens Material | Polycarbonate |