Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bovine Albumin Fraction V 0.2%

Catalog No. r08194
Click to view available options
Quantity:
20/Pk.
Each
Format:
F - 10mL
PB - 50mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Remel Bovine Albumin Fraction V 0.2% is used in the digestion-decontamination procedure for Mycobacterium spp.

Thermo Scientific™ Remel Bovine Albumin Fraction V 0.2% is used in the digestion-decontamination procedure for Mycobacterium spp.

TRUSTED_SUSTAINABILITY

Specifications

Description Bovine Albumin Fraction V 0.2%
Format F - 10mL
Product Type Buffer Solution
Quantity 20/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.