Learn More
Bovine Albumin Rabbit anti-Bovine, HRP, Polyclonal, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsDescription
Specifications
Specifications
| Antigen | Bovine Albumin |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Concentration | Total Protein: 28.4 mg/mL; Titer: 1:110;000 |
| Conjugate | HRP |
| Host Species | Rabbit |
| Quantity | 2 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Bovine |
| Show More |
Safety and Handling
For research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.