Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Bovine Beta Lactoglobulin ELISA Kit (Colorimetric)
SDP

Catalog No. NB599551KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB599551KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB599551KT Supplier Novus Biologicals™ Supplier No. NBP2599551KIT
Add to Cart
Add to Cart

High sensitivity and specificity for detection of bovine LGB.

The Bovine Beta Lactoglobulin ELISA Kit (Colorimetric) measures bovine Beta Lactoglobulin in serum, plasma and milk.

Specifications

Assay Range 0.156 to 10μg/mL
Assay Sensitivity 0.129μg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Milk, Plasma, Serum
Specificity For detection of bovine LGB.
Target Species Bovine
For Use With (Application) ELISA
Kit Contents Assay plate, Biotin-conjugate (100 x concentrate), Biotin-conjugate Diluent, Adhesive Strip (For 96 wells), HRP-avidin (100 x concentrate), HRP-avidin Diluent, Wash Buffer (25 x concentrate), TMB Substrate , Standard (Freeze dried), Sample Diluent ,Stop Solution and Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Beta Lactoglobulin
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.