Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine Coronavirus Monoclonal Antibody (5A4)
GREENER_CHOICE

Catalog No. PIMA123055
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA123055 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA123055 Supplier Invitrogen™ Supplier No. MA123055
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Bovine Coronavirus (BCoV) is a betacoronavirus affecting cattle, associated with both gastrointestinal and respiratory diseases. The gastrointestinal form causes neonatal calf diarrhea (calf scours) and winter dysentery in adult cattle, while the respiratory form contributes to bovine respiratory disease complex (shipping fever) affecting cattle of all ages. BCoVs exist as quasispecies without distinct genetic or antigenic markers distinguishing the clinical syndromes they cause. The virus is characterized by high morbidity but low mortality, with diagnosis typically based on serological or molecular methods. There is no registered antiviral treatment, so management relies on supportive therapy and preventative measures. BCoVs were also used as model viruses to study the survival potential of SARS-CoV-2 on various surfaces.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine Coronavirus
Applications ELISA
Classification Monoclonal
Clone 5A4
Concentration 5.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias BCV; Calf Enteritis; Winter Dysentery
Host Species Mouse
Immunogen Bovine corona virus.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.