Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Bovine Coronavirus Spike Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117562
Description
This antibody has been tested in direct-ELISA.
S1 attaches the virion to the cell membrane by binding to 9-O-acetylated sialic acid containing proteins, initiating the infection.
Specifications
| Bovine Coronavirus Spike Protein | |
| Polyclonal | |
| Unconjugated | |
| BCV Spike Protein; S glycoprotein; Spike glycoprotein | |
| Recombinant Bovine coronavirus Spike glycoprotein protein (326-540aa). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 2.86 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction