Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bovine IFN-gamma Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC2300U100UG
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Bovine samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IFN-gamma | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_776511 | |
IFNG | |
E. coli-derived recombinant bovine IFN-gamma Gln24-Thr166 Accession # NP_776511 | |
RUO | |
3458 | |
Bovine | |
IgG2a |
Flow Cytometry | |
345025 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Rat | |
100 μg | |
Primary | |
Detects bovine IFN-gamma in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with IFN-gamma from equine, canine, or feline systems is observed and no cross-reactivity with human, cotton rat, mouse, porcine, or rat IFN-gamma is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction