Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bovine IFN-gamma Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $585.50
Specifications
Antigen | IFN-gamma |
---|---|
Clone | 345001 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB23001SP
![]() |
R&D Systems
MAB23001SP |
25 μg |
Each for $200.50
|
|
|||||
MAB23001
![]() |
R&D Systems
MAB23001 |
500 μg |
Each for $585.50
|
|
|||||
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Bovine samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
IFN-gamma | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
IFNG | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
345001 | |
ELISA | |
Unconjugated | |
Rat | |
Bovine | |
NP_776511 | |
3458 | |
E. coli-derived recombinant bovine IFN-gamma Gln24-Thr166 Accession # NP_776511 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects bovine IFN-gamma in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant IFN-gamma from human, mouse or porcine is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title