Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine IFN-gamma Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2300
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2300 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2300 Supplier R&D Systems Supplier No. BAF2300
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IFN-gamma Polyclonal specifically detects IFN-gamma in Bovine samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen IFN-gamma
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. NP_776511
Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Goat
Immunogen E. coli-derived recombinant bovine IFN-gamma Gln24-Thr166 Accession # NP_776511
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3458
Test Specificity Detects bovine IFN-gamma in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human IFN-gamma , recombinant mouse IFN-gamma , and recombinant porcine IFN-gamma is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Bovine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.