Learn More
Description
Specifications
Specifications
| Antigen | IgG |
| Applications | Immunoprecipitation |
| Conjugate | Agarose |
| Dilution | Immunoprecipitation 1:10 - 1:500 |
| Formulation | 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Bovine |
| Purification Method | Multi-step |
| Quantity | 10 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
