Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine IgG Isotype Control, Alkaline Phosphatase, Novus Biologicals™
SDP

Catalog No. NBP197093 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197093 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197093 Supplier Novus Biologicals Supplier No. NBP197093
Add to Cart
Add to Cart

Bovine Antibody

IgG specifically detects IgG in Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IgG
Conjugate Alkaline Phosphatase (AP)
Formulation 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Bovine
Purification Method Multi-step
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Gene ID (Entrez) 3500
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Bovine IgG, anti-Bovine Serum and anti-Alkaline Phosphatase (calf intestine).
Target Species Bovine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.