Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine IL-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2469SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2469SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2469SP Supplier R&D Systems Supplier No. AF2469SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-4 Polyclonal specifically detects IL-4 in Bovine samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen IL-4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 10-20 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P30367
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbols IL4
Host Species Goat
Immunogen E. coli-derived recombinant bovine IL-4 His25-Cys135 Accession # P30367
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity Detects bovine IL-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant porcine IL-4 is observed, less than 5% cross-reactivity with recombinant human IL-4, recombinant feline IL-4, and recombinant canine IL-4 is observed, and less than 1% cross-reactivity with recombinant mouse IL-4, recombinant rat IL-4, and recombinant rhesus macaque IL-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Bovine
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.