Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bovine Lactoferrin Rabbit anti-Bovine, Ovine, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc BLAC101AP

Description
Bovine Lactoferrin Polyclonal antibody specifically detects Bovine Lactoferrin in Bovine, Ovine samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western BlotSpecifications
Bovine Lactoferrin | |
Polyclonal | |
Unconjugated | |
LTF | |
Bovine Lactoferrin; CKRX; Csp82; epididymis luminal protein 110; GIG12; Growth-inhibiting protein 12; HEL110; HLF2; Kaliocin 1; Kaliocin1; kaliocin-1; lactoferricin; Lactoferricin H; Lactoferricin-B; Lactoferricin-H; Lactoferrin; lactoferroxin; LactoferroxinA; Lactoferroxin-A; LactoferroxinB; Lactoferroxin-B; LactoferroxinC; Lactoferroxin-C; Lactotransferrin; LF; Lfcin H; Lfcin-B; LfcinH; Lfcin-H; LTF; minisatellite 10r detected by probe MMS10; MMS10R; Ms10r; N lobe; neutrophil lactoferrin; talalactoferrin | |
Purified Bovine Lactoferrin protein. | |
200 μL | |
Primary | |
Bovine, Ovine | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P24627 | |
Rabbit | |
Affinity chromatography | |
RUO | |
280846, 554321 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction