Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine Papilloma Virus Monoclonal Antibody (I24F)
GREENER_CHOICE

Catalog No. PIMA17407
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17407 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17407 Supplier Invitrogen™ Supplier No. MA17407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7407 antibody reacts with Bovine Papillomavirus in viral and infected tissue samples. MA1-7407 has been successfully used in ELISA applications. The MA1-7407 antibody was raised against a BPV preparation.

Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine Papilloma Virus
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone I24F
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias BPV
Host Species Mouse
Immunogen BPV preparation.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.