Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody (1E4)
GREENER_CHOICE

Catalog No. MA171535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA171535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA171535 Supplier Invitrogen™ Supplier No. MA171535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-71535 detects BPV-1 E2 protein in viral/infected samples. MA1-71535 has been successfully used in Western Blot and Immunofluorescence assays. The immunogen used to produce MA1-71535 is from amino acid region 250-280.

Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine Papilloma Virus Type 1 E2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative
Gene E2
Gene Accession No. P03122
Gene Alias BPV; BPV Protein E2; BPV-1; BPV1-E2
Gene Symbols E2
Host Species Mouse
Immunogen BPV-1 E2 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489020
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.