Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody (3F12)
GREENER_CHOICE

Catalog No. MA171533
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA171533 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA171533 Supplier Invitrogen™ Supplier No. MA171533
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody detects BPV type 1 E2 protein, amino acids 197-208 (GVSSTSSDFRDR).

Papillomaviruses are a diverse group of DNA-based viruses that infect the skin and mucous membranes of humans and a variety of animals. 100 different HPV types have been identified to date. Various types of HPV may cause warts while others may cause a subclinical infection resulting in precancerous lesions. All HPVs are transmitted by skin-to-skin contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bovine Papilloma Virus Type 1 E2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3F12
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and no preservative
Gene E2
Gene Accession No. P03122
Gene Alias BPV; BPV Protein E2; BPV-1; BPV1-E2
Gene Symbols E2
Host Species Mouse
Immunogen Recombinant full length BPV type 1 E2 protein purified from E. coli.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489020
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.